missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2L3 Protein
A cDNA sequence encoding the UBE2L3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13863-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBE2L3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAASRRLMKE LEEIRKCGMK NFRNIQVDEA NLLTWQGLIV PDNPPYDKGA FRIEINFPAE YPFKPPKITF KTKIYHPNID EKGQVCLPVI SAENWKPATK TDQVIQSLIA LVNDPQPEHP LRADLAEEYS KDRKKFCKNA EEFTKKYGEK RPVD | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein contains 50mM HEPES (pH 7.5) 150mM NaCl, 1mM DTT, and 10% glycerol. |