missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UBE2B Protein
A cDNA sequence encoding the UBE2B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13853-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
UBE2B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MHHHHHHAMGQLRSMSTPARRRLMRDFKRLQEDPPVGVSGAPSENNIMQWNAVIFGPEGTPFEDGTFKLVIEFSEEYPNKPPTVRFLSKMFHPNVYADGSICLDILQNRWSPTYDVSSILTSIQSLLDEPNPNSPANSQAAQLYQENKREYEKRVSAIVEQSWNDS | |
Greater than 95.0% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Lyophilized from a 0.2?m filtered concentrated (1 mg/ml) solution in 1X PBS and 1mM DTT, pH 7.5. |