missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Type-2 angiotensin II receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Type-2 angiotensin II receptor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5625-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000677.2 | |
Type-2 angiotensin II receptor Protein | |
Research Use Only | |
Agtr2 | |
Recombinant Protein | |
E. coli | |
MKGNSTLATT SKNITSGLHF GLVNISGNNE STLNCSQKPS DKHLD | |
Greater than 90% as determined by SDS-PAGE. |
186 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |