missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tubulin beta-6 chain Protein
A DNA sequence encoding the Homo sapiens (Human) Tubulin beta-6 chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8163-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001290453.1 | |
Tubulin beta-6 chain Protein | |
Research Use Only | |
TUBB6 | |
Recombinant Protein | |
E. coli | |
MREIVHIQAG QCGNQIGTKF WEVISDEHGI DPAGGYVGDS ALQLERINVY YNESSSQKYV PRAALVDLEP GTMDSVRSGP FGQLFRPDNF IFGQTGAGNN WAKGHYTEGA ELVDAVLDVV RKECEHCDCL QGFQLTHSLG GGTGSGMGTL LISKIREEFP DRIMNTFSVM PSPKVSDTVV EPYNATLSVH QLVENTDETY CIDNEALYDI CFRTLKLTTP TYGDLNHLVS ATMSGVTTSL RFPGQLNADL RKLAVNMVPF PRLHFFMPGF APLTSRGSQQ YRALTVPELT QQMFDARNMM AACDPRHGRY LTVATVFRGP MSMKEVDEQM LAIQSKNSSY FVEWIPNNVK VAVCDIPPRG LKMASTFIGN STAIQELFKR ISEQFSAMFR RKAFLHWFTG EGMDEMEFTE AESNMNDLVS EYQQYQDATA NDGEEAFEDE EEEIDG | |
Greater than 90% as determined by SDS-PAGE. |
84617 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |