missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSG Protein
A cDNA sequence encoding the TSG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10900-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
57045 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as measured by its ability to inhibit alkaline phosphatase production induced by rHuBMP-6 in mouse ATDC5 cells, is less than 16microg/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in PBS. |
TSG Protein | |
Research Use Only | |
TWSG1 | |
Recombinant Protein | |
Human | |
CNKALCASDV SKCLIQELCQ CRPGEGNCSC CKECMLCLGA LWDECCDCVG MCNPRNYSDT PPTSKSTVEE LHEPIPSLFR ALTEGDTQLN WNIVSFPVAE ELSHHENLVS FLETVNQPHH QNVSVPSNNV HAPYSSDKEH MCTVVYFDDC MSIHQCKISC ESMGASKYRW FHNACCECIG PECIDYGSKT VKCMN CMF | |
Greater than 95.0% as determined by SDS-PAGE. |