missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tryptophan 5-hydroxylase 2 Protein
A DNA sequence encoding the Homo sapiens(Human) Tryptophan 5-hydroxylase 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10004-iv-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_775489.2 | |
Tryptophan 5-hydroxylase 2 Protein | |
Research Use Only | |
TPH2 | |
Recombinant Protein | |
E. coli | |
MQPAMMMFSS KYWARRGFSL DSAVPEEHQL LGSSTLNKPN SGKNDDKGNK GSSKREAATE SGKTAVVFSL KNEVGGLVKA LRLFQEKRVN MVHIESRKSR RRSSEVEIFV DCECGKTEFN ELIQLLKFQT TIVTLNPPEN IWTEEEELED VPWFPRKISE LDKCSHRVLM YGSELDADHP GFKDNVYRQR RKYFVDVAMG YKYGQPIPRV EYTEEETKTW GVVFRELSKL YPTHACREYL KNFPLLTKYC GYREDNVPQL EDVSMFLKER SGFTVRPVAG YLSPRDFLAG LAYRVFHCTQ YIRHGSDPLY TPEPDTCHEL LGHVPLLADP KFAQFSQEIG LASLGASDED VQKLATCYFF TIEFGLCKQE GQLRAYGAGL LSSIGELKHA LSDKACVKAF DPKTTCLQEC LITTFQEAYF VSESFEEAKE KMRDFAKSIT RPFSVYFNPY TQSIEILKDT RSIENVVQDL RSDLNTVCDA LNKMNQYLGI | |
Greater than 85% as determined by SDS-PAGE. |
121278 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |