missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFSF8 Protein
A cDNA sequence encoding the TNFSF8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13773-HIS-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
944 | |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQRTDSIP NSPDNVPLKG GNCSEDLLCI LKRAPFKKSW AYLQVAKHLN KTKLSWNKDG ILHGVRYQDG NLVIQFPGLY FIICQLQFLV QCPNNSVDLK LELLINKHIK KQALVTVCES GMQTKHVYQN LSQFLLDYLQ VNTTISVNVD TFQYIDTSTF PLENVLSIFL YSNSD | |
Greater than 80% as determined by SDS-PAGE. |
TNFSF8 Protein | |
Research Use Only | |
TNFSF8 | |
Human | |
His | |
TNFSF8 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |