missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thymosin beta-4 Protein

A DNA sequence encoding the Human Thymosin beta-4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10182-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923008

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Especificaciones

Especificaciones

NP_066932.1
Thymosin beta-4 Protein
Research Use Only
TMSB4X
The biological activity determined by its ability to produce a protective effect against hydrogen peroxide in primary lung fibroblasts is in a concentration range of 0.5 - 10 ug/ml.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized
7114
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SDKPDMAEIE KFDKSKLKKT ETQEKNPLPS KETIEQEKQA GES
Greater than 97% as determined by SDS-PAGE and HPLC.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales