missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFB3 Protein
A DNA sequence encoding the Homo sapiens (Human) TGFB3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8903-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001316867.1 | |
TGFB3 Protein | |
Research Use Only | |
TGFB3 | |
Recombinant Protein | |
E. coli | |
ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS | |
Greater than 90% as determined by SDS-PAGE. |
7043 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |