missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-cell receptor alpha chain C region Protein
A DNA sequence encoding the Homo sapiens (Human) T-cell receptor alpha chain C region, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6836-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
T-cell receptor alpha chain C region Protein | |
Research Use Only | |
TRAC | |
Recombinant Protein | |
E. coli | |
PNIQNPDPAV YQLRDSKSSD KSVCLFTDFD SQTNVSQSKD SDVYITDKTV LDMRSMDFKS NSAVAWSNKS DFACANAFNN SIIPEDTFFP SPESSCDVKL VEKSFETDTN LNFQNLSVIG FRILLLKVAG FNLLMTLRLW SS | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |