missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUFU Protein
A cDNA sequence encoding the SUFU was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13639-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SUFU Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAELRPSGAP GPTAPPAPGP TAPPAFASLF PPGLHAIYGE CRRLYPDQPN PLQVTAIVKY WLGGPDPLDY VSMYRNVGSP SANIPEHWHY ISFGLSDLYG DNRVHEFTGT DGPSGFGFEL TFRLKRETGE SAPPTWPAEL MQGLARYVFQ SENTFCSGDH VSWHSPLDNS ESRIQHMLLT EDPQMQPVQT PFGVVTFLQI VGVCTEELHS AQQWNGQGIL ELLRTVPIAG GPWLITDMRR GETIFEIDPH LQERVDKGIE TDGSNLSGVS AKCAWDDLSR PPEDDEDSRS ICIGTQPRRL SGKDTEQIRE TLRRGLEINS KPVLPPINPQ RQNGLAHDRA PSRKDSLESD SSTAIIPHEL IRTRQLESVH LKFNQESGAL IPLCLRGRLL HGRHFTYKSI TGDMAITFVS TGVEGAFATE EHPYAAHGPW LQILLTEEFV EKMLEDLEDL TSPEEFKLPK EYSWPEKKLK VSILPDVVFD SPLH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SUFU protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 10% glycerol and 50mM NaCl. |