missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPECC1 Protein
A cDNA sequence encoding the SPECC1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13573-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SPECC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTQEGKII ELEQKCTGIL EQGRFEREKL LNIQQQLTCS LRKVEEENQG ALEMIKRLKE ENEKLNEFLE LERHNNNMMA KTLEECRVTL EGLKMENGSL KSHLQGEKQK ATEASAVEQT AESCEVQEML KVARAEKDLL ELSCNELRQE LLKANGEIKH VSSLLAKVEK DYSYLKEICD HQAEQLSRTS LKLQEKASES DAEIKDMKET IFELEDQVEQ HRAVKLHNNQ LISELESSVI KLEEQKSDLE RQLKTLTKQM KEETEEWRRF QADLQTAVVV ANDIKCEAQQ ELRTVKRKLL EEEEKNA | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SPECC1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol 1mM DTT and 0.1M NaCl. |