missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sodium / potassium-transporting ATPase subunit gamma Protein
A DNA sequence encoding the Homo sapiens (Human) Sodium/potassium-transporting ATPase subunit gamma, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6060-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001671.2 | |
Sodium / potassium-transporting ATPase subunit gamma Protein | |
Research Use Only | |
FXYD2 | |
Recombinant Protein | |
E. coli | |
MDRWYLGGSP KGDVDPFYYD YETVRNGGLI FAGLAFIVGL LILLSRRFRC GGNKKRRQIN EDEP | |
Greater than 80% as determined by SDS-PAGE. |
486 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |