missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Signal recognition particle 19 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Signal recognition particle 19 kDa protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6728-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001191122.1 | |
Signal recognition particle 19 kDa Protein | |
Research Use Only | |
SRP19 | |
Recombinant Protein | |
E. coli | |
ACAAARSPAD QDRFICIYPA YLNNKKTIAE GRRIPISKAV ENPTATEIQD VCSAVGLNVF LEKNKMYSRE WNRDVQYRGR VRVQLKQEDG SLCLVQFPSR KSVMLYAAEM IPKLKTRTQK TGGADQSLQQ GEGSKKGKGK KKK | |
Greater than 90% as determined by SDS-PAGE. |
6728 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |