missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SET Protein
A cDNA sequence encoding the SET was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13472-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SET Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPKRQS PLPPQKKKPR PPPALGPEET SASAGLPKKG EKEQQEAIEH IDEVQNEIDR LNEQASEEIL KVEQKYNKLR QPFFQKRSEL IAKIPNFWVT TFVNHPQVSA LLGEEDEEAL HYLTRVEVTE FEDIKSGYRI DFYFDENPYF ENKVLSKEFH LNESGDPSSK STEIKWKSGK DLTKRSSQTQ NKASRKRQHE EPESFFTWFT DHSDAGADEL GEVIKDDIWP NPLQYYLVPD MDDEEGEGEE DDDDDEEEEG LEDIDEEGDE DEGEEDEDDD EGEEGEEDEG EDD | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SET solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |