missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SERPINA3 Protein
A cDNA sequence encoding the SERPINA3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13456-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
SERPINA3 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MERMLPLLAL GLLAAGFCPA VLCHPNSPLD EENLTQENQD RGTHVDLGLA SANVDFAFSL YKQLVLKAPD KNVIFSPLSI STALAFLSLG AHNTTLTEIL KGLKFNLTET SEAEIHQSFQ HLLRTLNQSS DELQLSMGNA MFVKEQLSLL DRFTEDAKRL YGSEAFATDF QDSAAAKKLI NDYVKNGTRG KITDLIKDLD SQTMMVLVNY IFFKAKWEMP FDPQDTHQSR FYLSKKKWVM VPMMSLHHLT IPYFRDEELS CTVVELKYTG NASALFILPD QDKMEEVEAM LLPETLKRWR DSLEFREIGE LYLPKFSISR DYNLNDILLQ LGIEEAFTSK ADLSGITGAR NLAVSQVVHK AVLDVFEEGT EASAATAVKI TLLSALVETR TIVRFNRPFL MIIVPTDTQN IFFMSKVTNP KQA | |
Greater than 99.0%. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Lyophilized from 0.02M NH4HCO3. |