missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SEC61B Protein
A cDNA sequence encoding the SEC61B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13447-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
10952 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPGPTPS GTNVGSSGRS PSKAVAARAA GSTVRQRKNA SCGTRSAGRT TSAGTGGMWR FYTEDSPGLK VGP | |
Greater than 85.0% as determined by SDS-PAGE. |
SEC61B Protein | |
Research Use Only | |
SEC61B | |
Human | |
His | |
SEC61B protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |