missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAP30-binding Protein
A DNA sequence encoding the Homo sapiens (Human) SAP30-binding protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8186-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001288768.1 | |
SAP30-binding Protein | |
Research Use Only | |
SAP30BP | |
Recombinant Protein | |
E. coli | |
MAGKKNVLSS LAVYAEDSEP ESDGEAGIEA VGSAAEEKGG LVSDAYGEDD FSRLGGDEDG YEEEEDENSR QSEDDDSETE KPEADDPKDN TEAEKRDPQE LVASFSERVR NMSPDEIKIP PEPPGRCSNH LQDKIQKLYE RKIKEGMDMN YIIQRKKEFR NPSIYEKLIQ FCAIDELGTN YPKDMFDPHG WSEDSYYEAL AKAQKIEMDK LEKAKKERTK IEFVTGTKKG TTTNATSTTT TTASTAVADA QKRKSKWDSA IPVTTIAQPT ILTTTATLPA VVTVTTSASG SKTTVISAVG TIVKKAKQ | |
Greater than 80% as determined by SDS-PAGE. |
29115 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |