missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SAE1 Protein
A DNA sequence encoding the Homo sapiens (Human) SAE1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8183-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Specifica
NP_001139185.1 | |
SAE1 Protein | |
Research Use Only | |
SAE1 | |
Recombinant Protein | |
E. coli | |
MVEKEEAGGG ISEEEAAQYD RQIRLWGLEA QKRLRASRVL LVGLKGLGAE IAKNLILAGV KGLTMLDHEQ VTPEDPGAQF LIRTGSVGRN RAEASLERAQ NLNPMVDVKV DTEDIEKKPE SFFTQFDAVC LTCCSRDVIV KVDQICHKNS IKFFTGDVFG YHGYTFANLG EHEFVEEKTK VAKVSQGVED GPDTKRAKLD SSETTMVKKK VVFCPVKEAL EVDWSSEKAK AALKRTTSDY FLLQVLLKFR TDKGRDPSSD TYEEDSELLL QIRNDVLDSL GISPDLLPED FVRYCFSEMA PVCAVVGGIL AQEIVKALSQ RDPPHNNFFF FDGMKGNGIV ECLGPK | |
Greater than 80% as determined by SDS-PAGE. |
10055 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |