missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S-phase kinase-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) S-phase kinase-associated protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7528-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001230049.1 | |
S-phase kinase-associated protein 1 | |
Research Use Only | |
SKP2 | |
Recombinant Protein | |
E. coli | |
MHRKHLQEIP DLSSNVATSF TWGWDSSKTS ELLSGMGVSA LEKEEPDSEN IPQELLSNLG HPESPPRKRL KSKGSDKDFV IVRRPKLNRE NFPGVSWDSL PDELLLGIFS CLCLPELLKV SGVCKRWYRL ASDESLWQTL DLTGKNLHPD VTGRLLSQGV IAFRCPRSFM DQPLAEHFSP FRVQHMDLSN SVIEVSTLHG ILSQCSKLQN LSLEGLRLSD PIVNTLAKNS NLVRLNLSGC SGFSEFALQT LLSSCSRLDE LNLSWCFDFT EKHVQVAVAH VSETITQLNL SGYRKNLQKS DLSTLVRRCP NLVHLDLSDS VMLKNDCFQE FFQLNYLQHL SLSRCYDIIP ETLLELGEIP TLKTLQVFGI VPDGTLQLLK EALPHLQINC SHFTTIARPT IGNKKNQEIW GIKCRLTLQK PSCL | |
Greater than 90% as determined by SDS-PAGE. |
6502 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |