missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RuvB-like 2 Protein
A DNA sequence encoding the Homo sapiens (Human) RuvB-like 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8295-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001308120.1 | |
RuvB-like 2 Protein | |
Research Use Only | |
RUVBL2 | |
Recombinant Protein | |
E. coli | |
ATVTATTKVP EIRDVTRIER IGAHSHIRGL GLDDALEPRQ ASQGMVGQLA ARRAAGVVLE MIREGKIAGR AVLIAGQPGT GKTAIAMGMA QALGPDTPFT AIAGSEIFSL EMSKTEALTQ AFRRSIGVRI KEETEIIEGE VVEIQIDRPA TGTGSKVGKL TLKTTEMETI YDLGTKMIES LTKDKVQAGD VITIDKATGK ISKLGRSFTR ARDYDAMGSQ TKFVQCPDGE LQKRKEVVHT VSLHEIDVIN SRTQGFLALF SGDTGEIKSE VREQINAKVA EWREEGKAEI IPGVLFIDEV HMLDIESFSF LNRALESDMA PVLIMATNRG ITRIRGTSYQ SPHGIPIDLL DRLLIVSTTP YSEKDTKQIL RIRCEEEDVE MSEDAYTVLT RIGLETSLRY AIQLITAASL VCRKRKGTEV QVDDIKRVYS LFLDESRSTQ YMKEYQDAFL FNELKGETMD TS | |
Greater than 90% as determined by SDS-PAGE. |
10856 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |