missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS2 Protein
A cDNA sequence encoding the RPS2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13354-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RPS2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RPS2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),10% glycerol and 0.4M Urea. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADDAGA AGGPGGPGGP GMGNRGGFRG GFGSGIRGRG RGRGRGRGRG RGARGGKAED KEWMPVTKLG RLVKDMKIKS LEEIYLFSLP IKESEIIDFF LGASLKDEVL KIMPVQKQTR AGQRTRFKAF VAIGDYNGHV GLGVKCSKEV ATAIRGAIIL AKLSIVPVRR GYWGNKIGKP HTVPCKVTGR CGSVLVRLIP APRGTGIVSA PVPKKLLMMA GIDDCYTSAR GCTATLGNFA KATFDAISKT YSYLTPDLWK ETVFTKSPYQ EFTDHLVKTH TRVSVQRTQA PAVATT |