Learn More
enQuireBio™ Recombinant Human RELM-beta Protein
Recombinant Protein
Marca: enQuireBio™ QP5281-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RELM-beta Protein | |
Research Use Only | |
RELM-beta | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
25 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MQCSLDSVMD KKIKDVLNSL EYSPSPISKK LSCASVKSQG RPSSCPAGMA VTGCACGYGC GSWDVQLETT CHCQCSVVDW TTARCCHLT | |
Greater than 90 as determined by SDS-PAGE. |