Learn More
enQuireBio™ Recombinant Human Ragulator complex protein LAMTOR1 Protein
A DNA sequence encoding the Homo sapiens (Human) Ragulator complex protein LAMTOR1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7766-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_060377.1 | |
Ragulator complex protein LAMTOR1 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55004 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LAMTOR1 | |
Recombinant Protein | |
E. coli | |
MGCCYSSENE DSDQDREERK LLLDPSSPPT KALNGAEPNY HSLPSARTDE QALLSSILAK TASNIIDVSA ADSQGMEQHE YMDRARQYST RLAVLSSSLT HWKKLPPLPS LTSQPHQVLA SEPIPFSDLQ QVSRIAAYAY SALSQIRVDA KEELVVQFGI P |