missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RABIF Protein
A cDNA sequence encoding the RABIF was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13248-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RABIF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEPAEQPSELVSAEGRNRKA VLCQRCGSRV LQPGTALFSR RQLFLPSMRK KPALSDGSNP DGDLLQEHWL VEDMFIFENV GFTKDVGNIK FLVCADCEIG PIGWHCLDDK NSFYVALERV SHE | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RABIF protein solution (0.5 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol. |