missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPN7 Protein

A cDNA sequence encoding the PTPN7 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13212-1ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922859

Cantidad Precio
1 107.00€ / 1 microgramo
10 99.50€ / 1 microgramo
Fecha estimada de envío
07-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

PTPN7 Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PTPN7 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVQAHG GRSRAQPLTL SLGAAMTQPP PEKTPAKKHV RLQERRGSNV ALMLDVRSLG AVEPICSVNT PREVTLHFLR TAGHPLTRWA LQRQPPSPKQ LEEEFLKIPS NFVSPEDLDI PGHASKDRYK TILPNPQSRV CLGRAQSQED GDYINANYIR GYDGKEKVYI ATQGPMPNTV SDFWEMVWQE EVSLIVMLTQ LREGKEKCVH YWPTEEETYG PFQIRIQDMK ECPEYTVRQL TIQYQEERRS VKHILFSAWP DHQTPESAGP LLRLVAEVEE SPETAAHPGP IVVHCSAGIG RTGCFIATRI GCQQLKARGE VDILGIVCQL RLDRGGMIQT AEQYQFLHHT LALYAGQLPE EPSP
Videos
Documentos