missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTH Protein
A cDNA sequence encoding the PTH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10855-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PTH Protein | |
Research Use Only | |
The activity calculated by UMR106 cell/cAMP method corresponding to a specific activity of 10,000 Units/mg. | |
Human | |
Untagged | |
The protein (1 mg/ml) was lyophilized after extensive dialyses against 20mM PBS pH 7.0 and 150mM NaCl. |
500 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
LMHNLGKHLN SMERVEWLRK KLQDVHNF | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |