Learn More
enQuireBio™ Recombinant Human PTH Protein
Código de producto. p-7148665
Change view
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
500 microgramos
15995368 100 μg 100 microgramos
15915378 500 μg 500 microgramos
15905378 1 mg 1 miligramo
3 options
Código de producto. 15995368
Marca: enQuireBio™ QP10855100ug
A cDNA sequence encoding the PTH was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre PTH Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica The activity calculated by UMR106 cell/cAMP method corresponding to a specific activity of 10,000 Units/mg.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína Untagged
Secuencia LMHNLGKHLN SMERVEWLRK KLQDVHNF
Tampón The protein (1 mg/ml) was lyophilized after extensive dialyses against 20mM PBS pH 7.0 and 150mM NaCl.
Grado de pureza o calidad Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.