missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSPN Protein
A DNA sequence encoding the Human PSPN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10423-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004149.1 | |
PSPN Protein | |
Research Use Only | |
PSPN | |
The ED50 as determined by a cell proliferation assay using human TT medullary thyroid cancer cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
5623 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ALSGPCQLWS LTLSVAELGL GYASEEKVIF RYCAGSCPRG ARTQHGLALA RLQGQGRAHG GPCCRPTRYT DVAFLDDRHR WQRLPQLSAA ACGCGG | |
Greater than 97% as determined by SDS-PAGE and HPLC. |