missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSPN Protein

Product Code. p-7148226
Change view
Click to view available options
Cantidad:
100 μg
20 μg
250 μg
5 μg
Unit Size:
100 microgramos
20 microgramos
250 microgramos
5 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15963808 5 μg 5 microgramos
15943808 20 μg 20 microgramos
15933808 100 μg 100 microgramos
15953808 250 μg 250 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15963808 Supplier enQuireBio™ Supplier No. QP10423ec5ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human PSPN, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_004149.1
ID de gen (Entrez) 5623
Nombre PSPN Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Less than 1.0 EU/ug as determined by LAL method.
Símbolo de gen PSPN
Longitud de la proteína Partial (see sequence information for more details).
Actividad biológica The ED50 as determined by a cell proliferation assay using human TT medullary thyroid cancer cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína Untagged
Secuencia ALSGPCQLWS LTLSVAELGL GYASEEKVIF RYCAGSCPRG ARTQHGLALA RLQGQGRAHG GPCCRPTRYT DVAFLDDRHR WQRLPQLSAA ACGCGG
Tampón 0.2 um filtered PBS, pH 7.4, lyophilized
Grado de pureza o calidad Greater than 97% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.