missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMG3 Protein
A cDNA sequence encoding the PSMG3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13196-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PSMG3 Protein | |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PSMG3 solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 8.0) and 10% Glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEDTPLVISK QKTEVVCGVP TQVVCTAFSS HILVVVTQFG KMGTLVSLEP SSVASDVSKP VLTTKVLLGQ DEPLIHVFAK NLVAFVSQEA GNRAVLLAVA VKDKSMEGLK ALREVIRVCQ VW |