missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human pS2 / pNR-2 / Trefoil Factor 1 Protein
A DNA sequence encoding the Human pS2 / pNR-2 / Trefoil Factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10178-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_003216.1 | |
pS2 / pNR-2 / Trefoil Factor 1 Protein | |
Research Use Only | |
TFF1 | |
The ED50 as determined by a chemotaxis bioassay using human MCF-7 cells is less than 10 ug/ml, corresponding to a specific activity of > 100 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
7031 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
EAQTETCTVA PRERQNCGFP GVTPSQCANK GCCFDDTVRG VPWCFYPNTI DVPPEEECEF | |
Greater than 97% as determined by SDS-PAGE and HPLC. |