missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1B Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6520-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001028729.1 | |
Protein phosphatase 1B Protein | |
Research Use Only | |
PPM1B | |
Recombinant Protein | |
E. coli | |
SIVLVCFSNA PKVSDEAVKK DSELDKHLES RVEEIMEKSG EEGMPDLAHV MRILSAENIP NLPPGGGLAG KRNVIEAVYS RLNPHRESDG ASDEAEESGS QGKLVEALRQ MRINHRGNYR QLLEEMLTSY RLAKVEGEES PAEPAATATS SNSDAGNPVT MQESHTESES GLAELDSSNE DAGTKMSGEK I | |
Greater than 90% as determined by SDS-PAGE. |
5495 | |
50 μg | |
Not determined. | |
Full Length of Isoform Beta-X | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |