missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 1B Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8161-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001229393.1 | |
Protein phosphatase 1 regulatory subunit 1B Protein | |
Research Use Only | |
PPP1R1B | |
Recombinant Protein | |
E. coli | |
MLFRLSEHSS PEEEASPHQR ASGEGHHLKS KRPNPCAYTP PSLKAVQRIA ESHLQSISNL NENQASEEED ELGELRELGY PREEDEEEEE DDEEEEEEED SQAEVLKVIR QSAGQKTTCG QGLEGPWERP PPLDESERDG GSEDQVEDPA LSEPGEEPQR PSPSEPGT | |
Greater than 80% as determined by SDS-PAGE. |
84152 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |