missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 1B Protein

A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 1B, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8161-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16022981

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Especificaciones

Especificaciones

NP_001229393.1
Protein phosphatase 1 regulatory subunit 1B Protein
Research Use Only
PPP1R1B
Recombinant Protein
E. coli
MLFRLSEHSS PEEEASPHQR ASGEGHHLKS KRPNPCAYTP PSLKAVQRIA ESHLQSISNL NENQASEEED ELGELRELGY PREEDEEEEE DDEEEEEEED SQAEVLKVIR QSAGQKTTCG QGLEGPWERP PPLDESERDG GSEDQVEDPA LSEPGEEPQR PSPSEPGT
Greater than 80% as determined by SDS-PAGE.
84152
10 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions