missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein FAM160B1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein FAM160B1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7713-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001128523.1 | |
Protein FAM160B1 Protein | |
Research Use Only | |
FAM160B1 | |
Recombinant Protein | |
E. coli | |
MFSKFTSILQ HAVEALAPSL PLQEDFVYHW KAITHYYIET SDDKAPVTDT NIPSHLEQML DILVQEENER ESGETGPCME YLLHHKILET LYTLGKADCP PGMKQQVLVF YTKLLGRIRQ PLLPHINVHR PVQKLIRLCG EVLATPTENE EIQFLCIVCA KLKQDPYLVN FFLENKMKSL ASKGVPNVIS EDTLKGQDSL STDTGQSRQP EELSGATGME QTELEDEPPH QMDHLSTSLD NLSVTSLPEA SVVCPNQDYN LVNSLLNLTR SPDGRIAVKA CEGLMLLVSL PEPAAAKCLT QSTCLCELLT DRLASLYKAL PQSVDPLDIE TVEAINWGLD SYSHKEDASA FPGKRALISF LSWFDYCDQL IKEAQKTAAV ALAKAVHERF FIGVMEPQLM QTSEMGILTS TALLHRIVRQ VTSDVLLQEM VFFILGEQRE PETLAEISRH PLRHRLIEHC DHISDEISIM TLRMFEHLLQ KPNEHILYNL VLRNLEERNY TEYKPLCPED KDVVENGLIA GAVDLEEDPL FTDISPENTL PNQEWLSSSP PATPDHPKND GKTEVHKIVN SFLCLVPDDA KSSYHVEGTG YDTYLRDAHR QFRDYCAICL RWEWPGSPKA LEKCNLEAAF FEGHFLKVLF DRMGRILDQP YDVNLQVTSV LSRLSLFPHP HIHEYLLDPY VNLAPGCRSL FSVIVRVVGD LMLRIQRIQD FTPKLLLVRK RLLGLEPEGP MQTLPSWW | |
Greater than 90% as determined by SDS-PAGE. |
57700 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |