missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein atonal homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein atonal homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7932-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005163.1 | |
Protein atonal homolog 1 Protein | |
Research Use Only | |
ATOH1 | |
Recombinant Protein | |
Yeast | |
MSRLLHAEEW AEVKELGDHH RQPQPHHLPQ PPPPPQPPAT LQAREHPVYP PELSLLDSTD PRAWLAPTLQ GICTARAAQY LLHSPELGAS EAAAPRDEVD GRGELVRRSS GGASSSKSPG PVKVREQLCK LKGGVVVDEL GCSRQRAPSS KQVNGVQKQR RLAANARERR RMHGLNHAFD QLRNVIPSFN NDKKLSKYET LQMAQIYINA LSELLQTPSG GEQPPPPPAS CKSDHHHLRT AASYEGGAGN ATAAGAQQAS GGSQRPTPPG SCRTRFSAPA SAGGYSVQLD ALHFSTFEDS ALTAMMAQKN LSPSLPGSIL QPVQEENSKT SPRSHRSDGE FSPHSHYSDS DEAS | |
Greater than 90% as determined by SDS-PAGE. |
474 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |