missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Programmed cell death protein 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8043-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_009148.2 | |
Programmed cell death protein 10 Protein | |
Research Use Only | |
PDCD10 | |
Recombinant Protein | |
E. coli | |
MRMTMEEMKN EAETTSMVSM PLYAVMYPVF NELERVNLSA AQTLRAAFIK AEKENPGLTQ DIIMKILEKK SVEVNFTESL LRMAADDVEE YMIERPEPEF QDLNEKARAL KQILSKIPDE INDRVRFLQT IKDIASAIKE LLDTVNNVFK KYQYQNRRAL EHQKKEFVKY SKSFSDTLKT YFKDGKAINV FVSANRLIHQ TNLILQTFKT VA | |
Greater than 90% as determined by SDS-PAGE. |
11235 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |