missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PROC Protein
A cDNA sequence encoding the PROC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13151-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5624 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human (HEK293) | |
MWQLTSLLLFVATWGISGTPAPLDSVFSSSERAHQVLRIRKRANSFLEELRHSSLERE CIEEICDFEEAKEIFQNVDDTLAFWSKHVDGDQCLVLPLEHPCASLCCGHGTCIDGIG SFSCDCRSGWEGRFCQREVSFLNCSLDNGGCTHYCLEEVGWRRCSCAPGYKLGD DLLQCHPAVKFPCGRPWKRMEKKRSHLKRDTEDQEDQVDPRLIDGKMTRRGDSPW QVVLLDSKKKLACGAVLIHPSWVLTAAHCMDESKKLLVRLGEYDLRRWEKWELDLDIK EVFVHPNYSKSTTDNDIALLHLAQPATLSQTIVPICLPDSGLAERELNQAGQETLVTGW GYHSSREKEAKRNRTFVLNFIKIPVVPHNECSEVMSNMVSENMLCAGILGDRQDACE GDSGGPMVASFHGTWFLVGLVSWGEGCGLLHNYGVYTKVSRYLDWIHGHIRDKEAP QKSWAPEFHHHHHHHHThe first 32 amino acids are predicted as signal peptide | |
Greater than 80.0% as determined by SDS-PAGE. |
PROC Protein | |
Research Use Only | |
PROC | |
Human | |
His | |
PROC protein is supplied in 50mM Tris pH 7.5, 300mM NaCl and 10% Glycerol. |