missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable ATP-dependent RNA helicase DDX58 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable ATP-dependent RNA helicase DDX58, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5922-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_055129.2 | |
Probable ATP-dependent RNA helicase DDX58 Protein | |
Research Use Only | |
DDX58 | |
Recombinant Protein | |
E. coli | |
MTTEQRRSLQ AFQDYIRKTL DPTYILSYMA PWFREEEVQY IQAEKNNKGP MEAATLFLKF LLELQEEGWF RGFLDALDHA GYSGLYEAIE SWDFKKIEKL EEYRLLLKRL QPEFKTRIIP TDIISDLSEC LINQECEEIL QICSTKGMMA GAEKLVECLL RSDKENWPKT LKLALEKERN KFSELWIVEK GIKDVETEDL EDKMETSDIQ IFYQEDPECQ NLSENSCPPS EVSDTNLYSP FKPRNYQLEL ALPAMKGKNT IICAPTGCGK TFVSLLICEH HLKKFPQGQK GKVVFFANQI PVYEQQKSVF SKYFERHGYR VTGISGATAE NVPVEQIVEN NDIIILTPQI LVNNLKKGTI PSLSIFTLMI FDECHNTSKQ HPYNMIMFNY LDQKLGGSSG PLPQVIGLTA SVGVGDAKNT DEALDYICKL | |
Greater than 90% as determined by SDS-PAGE. |
23586 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |