Learn More
enQuireBio™ Recombinant Human PRKRA Protein
A cDNA sequence encoding the PRKRA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13146-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PRKRA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSQSRHR AEAPPLERED SGTFSLGKMI TAKPGKTPIQ VLHEYGMKTK NIPVYECERS DVQIHVPTFT FRVTVGDITC TGEGTSKKLA KHRAAEAAIN ILKANASICF AVPDPLMPDP SKQPKNQLNP IGSLQELAIH HGWRLPEYTL SQEGGPAHKR EYTTICRLES FMETGKGASK KQAKRNAAEK FLAKFSNISP ENHISLTNVV GHSLGCTWHS LRNSPGEKIN LLKRSLLSIP NTDYIQLLSE IAKEQGFNIT YLDIDELSAN GQYQCLAELS TSPITVCHGS GISCGNAQSD AAHNALQYLK IIAERK | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRKRA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA. |