missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKACA Protein
A cDNA sequence encoding the PRKACA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13140-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5566 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
PRKACA | |
Human | |
Untagged | |
PKA catalytic subunit a is supplied in a buffer containing 20mM MOPS pH 7, 150mM NaCl, 1mM DTT, 1mM EDTA and 50% Glycerin. |
PRKACA Protein | |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Insect | |
MGNAAAAKKG SEQESVKEFL AKAKEDFLKK WESPAQNTAH LDQFERIKTL GTGSFGRVML VKHKETGNHY AMKILDKQKV VKLKQIEHTL NEKRILQAVN FPFLVKLEFS FKDNSNLYMV MEYVPGGEMF SHLRRIGRFS EPHARFYAAQ IVLTFEYLHS LDLIYRDLKP ENLLIDQQGY IQVTDFGFAK RVKGRTWTLC GTPEYLAPEI ILSKGYNKAV DWWALGVLIY EMAAGYPPFF ADQPIQIYEK IVSGKVRFPS HFSSDLKDLL RNLLQVDLTK RFGNLKNGVN DIKNHKWFAT TDWIAIYQRK VEAPFIPKFK GPGDTSNFDD YEEEEIRVSI NEKCGKEFSE F |