missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRDX5 Protein

A cDNA sequence encoding the PRDX5 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10841-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15995328

Cantidad Precio
1 129.74€ / 5 microgramos
10 121.33€ / 5 microgramos
Envío estimado: 10-03-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Especificaciones

Especificaciones

PRDX5 Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MAPIKVGDAI PAVEVFEGEP GNKVNLAELF KGKKGVLFGV PGAFTPGCSK THLPGFVEQA EALKAKGVQV VACLSVNDAF VTGEWGRAHK AEGKVRLLAD PTGAFGKETD LLLDDSLVSI FGNRRLKRFS MVVQDGIVKA LNVEPDGTGL TCSLAPNIIS QL
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The specific activity was found to be approximately 117-136 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX5 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute.
Human
Untagged
The PRDX5 solution contains 20mM HEPES buffer (pH 7.4).
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales