missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRDX5 Protein
A cDNA sequence encoding the PRDX5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10841-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PRDX5 Protein | |
Research Use Only | |
The specific activity was found to be approximately 117-136 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX5 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute. | |
Human | |
Untagged | |
The PRDX5 solution contains 20mM HEPES buffer (pH 7.4). |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPIKVGDAI PAVEVFEGEP GNKVNLAELF KGKKGVLFGV PGAFTPGCSK THLPGFVEQA EALKAKGVQV VACLSVNDAF VTGEWGRAHK AEGKVRLLAD PTGAFGKETD LLLDDSLVSI FGNRRLKRFS MVVQDGIVKA LNVEPDGTGL TCSLAPNIIS QL | |
Greater than 95.0% as determined by SDS-PAGE. |