missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1CC Protein
A cDNA sequence encoding the PPP1CC was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10836-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5501 | |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 700 units/mg, and is defined as the amount of enzyme that hydrolyzes 1.0 nmole of p-nitrophenyl phosphate (pNPP) per minute at pH 7.5 at 37C. | |
Human | |
His | |
The PPP1CC protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 50% glycerol. |
PPP1CC Protein | |
Research Use Only | |
PPP1CC | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MADLDKLNID SIIQRLLEVR GSKPGKNVQL QENEIRGLCL KSREIFLSQP ILLELEAPLK ICGDIHGQYY DLLRLFEYGG FPPESNYLFL GDYVDRGKQS LETICLLLAY KIKYPENFFL LRGNHECASI NRIYGFYDEC KRRYNIKLWK TFTDCFNCLP IAAIVDEKIF CCHGGLSPDL QSMEQIRRIM RPTDVPDQGL LCDLLWSDPD KDVLGWGEND RGVSFTFGAE VVAKFLHKHD LDLICRAHQV VEDGYEFFAK RQLVTLFSAP NYCGEFDNAG AMMSVDETLM CSFQILKPAE KKKPNATRPV TPPRGMITKQ AKK | |
Greater than 85.0% as determined by SDS-PAGE. |