missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Poly(A)-specific ribonuclease PARN Protein
A DNA sequence encoding the Homo sapiens (Human) Poly(A)-specific ribonuclease PARN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6467-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001127949.1 | |
Poly(A)-specific ribonuclease PARN Protein | |
Research Use Only | |
PARN | |
Recombinant Protein | |
Yeast | |
MEIIRSNFKS NLHKVYQAIE EADFFAIDGE FSGISDGPSV SALTNGFDTP EERYQKLKKH SMDFLLFQFG LCTFKYDYTD SKYITKSFNF YVFPKPFNRS SPDVKFVCQS SSIDFLASQG FDFNKVFRNG IPYLNQEEER QLREQYDEKR SQANGAGALS YVSPNTSKCP VTIPEDQKKF IDQVVEKIED LLQSEENKNL DLEPCTGFQR KLIYQTLSWK YPKGIHVETL ETEKKERYIV ISKVDEEERK RREQQKHAKE QEELNDAVGF SRVIHAIANS GKLVIGHNML LDVMHTVHQF YCPLPADLSE FKEMTTCVFP RLLDTKLMAS TQPFKDIINN TSLAELEKRL KETPFNPPKV ESAEGFPSYD TASEQLHEAG YDAYITGLCF ISMANYLGSF LSPPKIHVSA RSKLIEPFFN KLFLMRVMDI PYLNLEGPDL QPKRDHVLHV TFPKEWKTSD LYQLFSAFGN IQISWIDDTS AFVSLSQPEQ VKIAVNTSKY AESYRIQTYA EYMGRKQEEK QIKRKWTEDS WKEADSKRLN PQCIPYTLQN HYYRNNSFTA PSTVGKRNLS PSQEEAGLED GVSGEISDTE LEQTDSCAEP LSEGRKKAKK LKRMKKELSP AGSISKNSPA TLFEVPDTW | |
Greater than 90% as determined by SDS-PAGE. |
5073 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |