missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLK1 / PLK-1 Protein
A DNA sequence encoding the Homo sapiens (Human) PLK1 / PLK-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10044-ba-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005021.2 | |
PLK1 / PLK-1 Protein | |
Research Use Only | |
PLK1 | |
Recombinant Protein | |
Baculovirus | |
MSAAVTAGKL ARAPADPGKA GVPGVAAPGA PAAAPPAKEI PEVLVDPRSR RRYVRGRFLG KGGFAKCFEI SDADTKEVFA GKIVPKSLLL KPHQREKMSM EISIHRSLAH QHVVGFHGFF EDNDFVFVVL ELCRRRSLLE LHKRRKALTE PEARYYLRQI VLGCQYLHRN RVIHRDLKLG NLFLNEDLEV KIGDFGLATK VEYDGERKKT LCGTPNYIAP EVLSKKGHSF EVDVWSIGCI MYTLLVGKPP FETSCLKETY LRIKKNEYSI PKHINPVAAS LIQKMLQTDP TARPTINELL NDEFFTSGYI PARLPITCLT IPPRFSIAPS SLDPSNRKPL TVLNKGLENP LPERPREKEE PVVRETGEVV DCHLSDMLQQ LHSVNASKPS ERGLVRQEEA EDPACIPIFW VSKWVDYSDK YGLGYQLCDN SVGVLFNDST RLILYNDGDS LQYIERDGTE SYLTVSSHPN SLMKKITLLK YFRNYMSEHL LKAGANITPR EGDELARLPY LRTWFRTRSA IILHLSNGSV QINFFQDHTK LILCPLMAAV TYIDEKRDFR TYRLSLLEEY GCCKELASRL RYARTMVDKL LSSRSASNRL KAS | |
Greater than 90% as determined by SDS-PAGE. |
5347 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |