missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pleiotrophin / PTN / HB-GAM Protein
Recombinant Protein
Marca: enQuireBio™ QP5365-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001308315.1 | |
Pleiotrophin / PTN / HB-GAM Protein | |
Research Use Only | |
PTN | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
5764 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGKKEKPEKK VKKSDCGEWQ WSVCVPTSGD CGLGTREGTR TGAECKQTMK TQRCKIPCNW KKQFGAECKY QFQAWGECDL NTALKTRTGS LKRALHNAEC QKTVTISKPC GKLTKPKPQA ESKKKKKEGK KQEKMLD | |
Greater than 95% as determined by SDS-PAGE. |