missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PKC-a Protein
A cDNA sequence encoding the PKC-a was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13064-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5578 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MADVYPGNDS TASQDVANRF ARKGALRQKN VHEVKDHKFI ARFFKQPTFC SHCTDFIWGF GKQGFQCQVC CFVVHKRCHE FVTFSCPGAD KGPDTDDPRS KHKFKIHTYG SPTFCDHCGS LLYGLIHQGM KCDTCDMNVH KQCVINVPSL CGMDHTEKRG RIYLKAEVAD EKLHVTVRDA KNLIPMDPNG LSDPYVKLKL IPDPKNESKQ KTKTIRSTLN PQWNESFTFK LKPSDKDRRL SVEIWDWDRT TRNDFMGSLS FGVSELMKMP ASGWYKLLNQ EEGEYYNVPI PEGDEEGNME LRQKFEKAKL GPAGNKVISP SEDRKQPSNN LDRVKLTDFN FLMVLGKGSF GKVMLADRKG TEELYAIKIL KKDVVIQDDD VECTMVEKRV LALLDKPPFL TQLHSCFQTV DRLYFVMEYV NGGDLMYHIQ QVGKFKEPQA VFYAAEISIG LFFLHKRGII YRDLKLDNVM LDSEGHIKIA DFGMCKEHMM DGVTTRTFCG TPDYIAPEII AYQPYGKSVD WWAYGVLLYE MLAGQPPFDG EDEDELFQSI MEHNVSYPKS LSKEAVSICK GLMTKHPAKR LGCGPEGERD VREHAFFRGI DWEKLENREI QPPFKPKVCG KGAENFDKFF TRGQPVLTPP DQLVIANIDQ SDFEGFSYVN PQFVHPILQS AV | |
Greater than 95% as determined by SDS-PAGE. |
PKC-a Protein | |
Research Use Only | |
PRKCA | |
Human | |
His | |
PKC-a is supplied in 20mM HEPES pH 7.5, 250mM NaCl, 50% glycerol, 2mM EDTA, 2mM EGTA, 0.05%Triton X-100 and 5mM DTT. |