missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PGAM1 Protein
A cDNA sequence encoding the PGAM1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10812-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PGAM1 Protein | |
Research Use Only | |
Specific activity is >300 units/mg, in which One unit will convert 1.0 umole of 3-phosphoglycerate to 2-phosphoglcerate per minute at pH 7.6 at 37C. | |
Human | |
His | |
The PGAM1 1 mg/ml protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol. |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAYKLVLIR HGESAWNLEN RFSGWYDADL SPAGHEEAKR GGQALRDAGY EFDICFTSVQ KRAIRTLWTV LDAIDQMWLP VVRTWRLNER HYGGLTGLNK AETAAKHGEA QVKIWRRSYD VPPPPMEPDH PFYSNISKDR RYADLTEDQL PSCESLKDTI ARALPFWNEE IVPQIKEGKR VLIAAHGNSL RGIVKHLEGL SEEAIMELNL PTGIPIVYEL DKNLKPIKPM QFLGDEETVR KAMEAVAAQG KAKK | |
Greater than 90% as determined by SDS-PAGE. |